accessibility menu, dialog, popup

UserName

CH3(R)(S)(R)H3C(R)O(R)OCH3(S)(R)OOOOHOO

CAS RN 88495-63-0

IUPAC Name: 4-oxo-4-{[(4S,5R,8S,9R,10R,12R,13R)-1,5,9-trimethyl-11,14,15,16-tetraoxatetracyclo[10.3.1.0⁴,¹³.0⁸,¹³]hexadecan-10-yl]oxy}butanoic acid
Synonyms: artesunate
Molecular Weight (g/mol): 384.43
Molecular Formula: C19H28O8
InChi Key: FIHJKUPKCHIPAT-JQXDXKTESA-N
SMILES: C[C@@H]1CC[C@H]2[C@@H](C)[C@@H](OC(=O)CCC(O)=O)O[C@@H]3OC4(C)CC[C@@H]1[C@@]23OO4
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)