missing translation for 'onlineSavingsMsg'
Learn More

Artesunate 98.0+%, TCI America™

Catalog No. p-7117783
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A21915G 5 g
A219125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A21915G Supplier TCI America Supplier No. A21915G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 88495-63-0
Molecular Formula C19H28O8
Molecular Weight (g/mol) 384.425
MDL Number MFCD00866204
InChI Key FIHJKUPKCHIPAT-RVBFJZMGSA-N
Synonym artesunate
PubChem CID 129317578
SMILES CC1CCC2C(C(OC3C24C1CCC(O3)(OO4)C)OC(=O)CCC(=O)O)C

Specifications

Melting Point 133°C
Color White
Quantity 5 g
Formula Weight 384.43
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Artesunate
TSCA No
Recommended Storage Refrigerator