missing translation for 'onlineSavingsMsg'
Learn More

Artemisinin 97.0+%, TCI America™

Catalog No. p-7117740
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A21181G 1 g
A21185G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A21181G Supplier TCI America Supplier No. A21181G
Only null left

Chemical Identifiers

CAS 63968-64-9
Molecular Formula C15H22O5
Molecular Weight (g/mol) 282.336
MDL Number MFCD00081057
InChI Key BLUAFEHZUWYNDE-DKGJTOOQSA-N
Synonym artemisinine, artemisinin, gnf-pf-5341, artemisinin, 18, 1s,4s,5r,8s,9r,12s,13r-1,5,9-trimethyl-11,14,15,16-tetraoxatetracyclo 10.3.1.0^ 4,13 .0^ 8,13 hexadecan-10-one, 1s,4s,5r,8s,9r,12s,13r-1,5,9-trimethyl-11,14,15,16-tetraoxatetracyclo 10.3.1.04,13.08,13 hexadecan-10-one
PubChem CID 9838675
SMILES CC1CCC2C(C(=O)OC3C24C1CCC(O3)(OO4)C)C

Specifications

Melting Point 153°C
Color White
Quantity 1 g
Formula Weight 282.34
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Artemisinin
RTECSNumber KD4170000
TSCA No