accessibility menu, dialog, popup

UserName

CAS RN 63968-64-9

IUPAC Name: (1R,4S,5R,8S,9R,12S,13R)-1,5,9-trimethyl-11,14,15,16-tetraoxatetracyclo[10.3.1.0⁴,¹³.0⁸,¹³]hexadecan-10-one
Synonyms: (+)-artemisinin qinghaosu artemisinin antimalarial extract artemisia annua L. qinghosu artemisine QHS artemisinine artemisinina quinghaosu
Molecular Weight (g/mol): 282.34
Molecular Formula: C15H22O5
InChi Key: BLUAFEHZUWYNDE-NNWCWBAJSA-N
SMILES: C[C@@H]1CC[C@H]2[C@@H](C)C(=O)O[C@@H]3O[C@@]4(C)CC[C@@H]1[C@@]23OO4
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)