missing translation for 'onlineSavingsMsg'
Learn More

Artemisinin, 98%

CAS: 63968-64-9 | C15H22O5 | 282.336 g/mol

Supplier:  Thermo Scientific Chemicals J6540606

 View more versions of this product

Catalog No. AAJ6540606

Only null left
Explore available promotions

Description

Description

Artemisinin inhibits angiogenesis by down-regulating HIF-1α and VEGF expression in mouse embryonic stem cells. Artemisinin crosses the blood-brain barrier and is an inhibitor of human NOS2 (iNOS).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble to 100mM in DMSO and to 75mM in ethanol

Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agents, heat. Store in refrigerator.
Specifications

Specifications

Artemisinin
156°C to 157°C
C15H22O5
5 g
artemisinine, artemisinin, gnf-pf-5341, artemisinin, 18, 1s,4s,5r,8s,9r,12s,13r-1,5,9-trimethyl-11,14,15,16-tetraoxatetracyclo 10.3.1.0^ 4,13 .0^ 8,13 hexadecan-10-one, 1s,4s,5r,8s,9r,12s,13r-1,5,9-trimethyl-11,14,15,16-tetraoxatetracyclo 10.3.1.04,13.08,13 hexadecan-10-one
BLUAFEHZUWYNDE-DKGJTOOQSA-N
CC1CCC2C(C(=O)OC3C24C1CCC(O3)(OO4)C)C
9838675
99%
63968-64-9
White
MFCD00081057
14,817
Soluble to 100mM in DMSO and to 75mM in ethanol
+76° (c=0.5 in Methanol)
282.336
282.33
Crystals or Powder

Chemical Identifiers

63968-64-9
282.336
BLUAFEHZUWYNDE-DKGJTOOQSA-N
9838675
C15H22O5
MFCD00081057
artemisinine, artemisinin, gnf-pf-5341, artemisinin, 18, 1s,4s,5r,8s,9r,12s,13r-1,5,9-trimethyl-11,14,15,16-tetraoxatetracyclo 10.3.1.0^ 4,13 .0^ 8,13 hexadecan-10-one, 1s,4s,5r,8s,9r,12s,13r-1,5,9-trimethyl-11,14,15,16-tetraoxatetracyclo 10.3.1.04,13.08,13 hexadecan-10-one
CC1CCC2C(C(=O)OC3C24C1CCC(O3)(OO4)C)C
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : KD4170000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only