missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Artemis Recombinant Rabbit Monoclonal Antibody (HL1227)
GREENER_CHOICE

Catalog No. pima546962
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA546962 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA546962 Supplier Invitrogen™ Supplier No. MA546962
Only null left

Rabbit Recombinant Monoclonal Antibody

Store as concentrated solution. Centrifuge briefly prior to opening vial.

The protein encoded by this gene is a receptor for aryl hydrocarbons and a ligand-activated transcription factor. The encoded protein is found in the cytoplasm as part of a multiprotein complex, but upon binding of ligand is transported to the nucleus. This protein can regulate the expression of many xenobiotic metabolizing enzymes. Also, the encoded protein can bind specifically to and inhibit the activity of hepatitis B virus.

Specifications

Antigen Artemis
Applications Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone HL1227
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene DCLRE1C
Gene Accession No. Q96SD1
Gene Alias 9930121L06Rik; AI661365; Art; ARTEMIS; ASCID; A-SCID; Dclre1c; DCLRE1C protein; DCLREC1C; DNA cross-link repair 1A, PSO2 homolog; DNA cross-link repair 1C; DNA cross-link repair 1C (PSO2 homolog, S. cerevisiae); DNA cross-link repair 1C protein; DNA cross-link repair 1C, PSO2 homolog (S. cerevisiae); FLJ11360; hSNM1C; mArt; Protein artemis; Protein A-SCID; RS SCID; RS-SCID; SCIDA; severe combined immunodeficiency, type a (Athabascan); SNM1 homolog C; SNM1C; SNM1C antibody; Snm1l; SNM1-like protein; tein
Gene Symbols DCLRE1C
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human Artemis. The exact sequence is proprietary.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64421
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less