missing translation for 'onlineSavingsMsg'
Learn More

Artemether, Thermo Scientific Chemicals

Catalog No. p-7214385
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC460700050 5 g
AC460700250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC460700050 Supplier Thermo Scientific Chemicals Supplier No. 460700050
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 71963-77-4
Molecular Formula C16H26O5
Molecular Weight (g/mol) 298.38
InChI Key SXYIRMFQILZOAM-HVNFFKDJSA-N
IUPAC Name (1R,4S,5R,8S,9R,10S,12R,13R)-10-methoxy-1,5,9-trimethyl-11,14,15,16-tetraoxatetracyclo[10.3.1.0⁴,¹³.0⁸,¹³]hexadecane
SMILES CO[C@H]1O[C@@H]2O[C@@]3(C)CC[C@H]4[C@H](C)CC[C@@H]([C@H]1C)[C@@]24OO3

Specifications

Melting Point 86.0°C to 89.0°C
Quantity 5 g
Formula Weight 298.37
Chemical Name or Material Artemether
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Harmful if swallowed.

Precautionary Statement GHS P Statement Wash face, hands and any exposed skin thoroughly after handling.

IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

RUO – Research Use Only