missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Artemether 98.0+%, TCI America™
$149.13 - $441.78
Chemical Identifiers
| CAS | 71963-77-4 |
|---|---|
| Molecular Formula | C16H26O5 |
| Molecular Weight (g/mol) | 298.379 |
| MDL Number | MFCD00866205 |
| InChI Key | SXYIRMFQILZOAM-ZSLLRAOYSA-N |
| PubChem CID | 132451910 |
| SMILES | CC1CCC2C(C(OC3C24C1CCC(O3)(OO4)C)OC)C |
Chemical Identifiers
| 71963-77-4 | |
| 298.379 | |
| SXYIRMFQILZOAM-ZSLLRAOYSA-N | |
| CC1CCC2C(C(OC3C24C1CCC(O3)(OO4)C)OC)C |
| C16H26O5 | |
| MFCD00866205 | |
| 132451910 |
Specifications
| 71963-77-4 | |
| White | |
| MFCD00866205 | |
| SXYIRMFQILZOAM-ZSLLRAOYSA-N | |
| 298.379 | |
| 298.38 | |
| Crystalline Powder |
| 89°C | |
| C16H26O5 | |
| 5 g | |
| CC1CCC2C(C(OC3C24C1CCC(O3)(OO4)C)OC)C | |
| 132451910 | |
| ≥98.0% (HPLC) | |
| Artemether |
Safety and Handling
RTECSNumber : KD4165000
TSCA : No