missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ART5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560610
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560610 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560610 Supplier Invitrogen™ Supplier No. PA560610
Only null left

Rabbit Polyclonal Antibody

Immunogen sequence: LQLLRGSGGC SRGPGEVVFR GVGSLRFEPK RLGDSVRLGQ FASSSLDKAV AHRFGNATLF SLTTCFGAPI QAFSV Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 83%.

The protein encoded by this gene belongs to the ARG-specific ADP-ribosyltransferase family. Proteins in this family regulate the function of target proteins by attaching ADP-ribose to specific amino acid residues in their target proteins. The mouse homolog lacks a glycosylphosphatidylinositol-anchor signal sequence and is predicted to be a secretory enzyme. Transcript variants with different 5' UTRs, but encoding the same protein have been found for this gene.

Specifications

Antigen ART5
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ART5
Gene Accession No. Q96L15
Gene Alias ADP-ribosyltransferase 5; ADP-ribosyltransferase C2 and C3 toxin-like 5; Art5; ARTC5; Ecto-ADP-ribosyltransferase 5; mono(ADP-ribosyl)transferase 5; NAD(P)(+)--arginine ADP-ribosyltransferase 5; UNQ575/PRO1137; YAC-2
Gene Symbols ART5
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ART5. Recombinant protein control fragment (Product #RP-97449).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116969
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less