missing translation for 'onlineSavingsMsg'
Learn More

Arsenic Polycrystalline lump, 2-8mm (0.08-0.3 in.), Puratronic™, 99.9999% (Metals basis), Thermo Scientific™

Catalog No. p-4824614
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
1 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1337509 10 g
AA1337514 25 g
AA1337522 100 g
AA13375A1 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AA1337509 Supplier Thermo Scientific Chemicals Supplier No. 1337509
Only null left

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 7440-38-2
Molecular Formula As
Molecular Weight (g/mol) 74.92
MDL Number MFCD00085309
InChI Key RQNWIZPPADIBDY-UHFFFAOYSA-N
Synonym arsenicals, arsenic, inorganic, arsenic-75, arsen, gray, grey, black, metallic, compounds, colloidal
PubChem CID 5359596
ChEBI CHEBI:27563
IUPAC Name arsenic
SMILES [As]

Specifications

Color Gray
Physical Form Polycrystalline Lump
Quantity 10 g
Assay Percent Range 99.9999% (Metals basis)
Odor Odorless
Appearance Gray lump
Chemical Name or Material Arsenic
Hazard Category H301+H331-H350
Precautionary Statement P201-P202-P261-P264b-P270-P271-P281-P301+P310-P304+P340-P308+P313-P311-P330-P501c
DOTInformation DOT Class: 6.1, Packaging Group: II
EINECSNumber 213-148-6
TSCA Yes
Recommended Storage Ambient temperatures