missing translation for 'onlineSavingsMsg'
Learn More

Arsenic(III) telluride, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog No. AA4170806
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4170806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA4170806 Supplier Thermo Scientific Chemicals Supplier No. 04170806

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 12044-54-1
Molecular Formula As2Te3
Molecular Weight (g/mol) 532.643
MDL Number MFCD00014167
InChI Key GTIUFDICMGTSPM-UHFFFAOYSA-N
Synonym arsenic iii telluride, tellanylidenearsanyl tellanyl arsanetellone, arsenic tritelluride
PubChem CID 15770922
SMILES [As](=[Te])[Te][As]=[Te]

Specifications

Melting Point 381°C
Physical Form Powder
Quantity 5 g
UN Number UN1557
Formula Weight 532.64
Percent Purity 99.999%
Odor Odorless
Density 6.50 g/mL
Assay (metals basis)
Chemical Name or Material Arsenic(III) telluride
Hazard Category H301+H331-H350
Hazard Statement GHS H Statement
H301-H331
Toxic if swallowed.
Toxic if inhaled.
Precautionary Statement P201-P202-P261-P264b-P270-P271-P281-P301+P310-P304+P340-P308+P313-P311-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ARSENIC COMPOUNDS, SOLID, N.O.S.
EINECSNumber 234-955-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only