accessibility menu, dialog, popup

UserName

AsAsTeTeTe

CAS RN 12044-54-1

IUPAC Name: diarsenic tritellurium
Synonyms: diarsenic tritellurium
Molecular Weight (g/mol): 532.64
Molecular Formula: As2Te3
InChi Key: KOOQXSDVIQPGQD-UHFFFAOYSA-N
SMILES: [As].[As].[Te].[Te].[Te]
Percent Purity 
  • (2)
Physical Form 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results