missing translation for 'onlineSavingsMsg'
Learn More

Arsenic(III) iodide, 99.999% (metals basis)

Catalog No. p-7051783
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4196206 5 g
AA4196214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4196206 Supplier Thermo Scientific Chemicals Supplier No. 04196206

CAS: 7784-45-4 | AsI3 | 455.635 g/mol

Arsenic(III) iodide is useful for preparing organoarsenic compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7784-45-4
Molecular Formula AsI3
Molecular Weight (g/mol) 455.635
MDL Number MFCD00014170
InChI Key IKIBSPLDJGAHPX-UHFFFAOYSA-N
Synonym arsenous triiodide, arsenic iii iodide, arsenic iodide, arsenic triiodide, triiodoarsine, arsenous iodide, hsdb 420, arsen iii-jodid
PubChem CID 24575
IUPAC Name triiodoarsane
SMILES [As](I)(I)I

Specifications

Boiling Point 424°C
Melting Point 140°C
Physical Form Powder
Quantity 5 g
UN Number UN1557
Merck Index 14,803
Solubility Information Hydrolyzes slowly in water.
Formula Weight 455.64
Percent Purity 99.999%
Odor Pungent
Sensitivity Moisture sensitive
Density 4.39 g/mL
Assay (metals basis)
Chemical Name or Material Arsenic(III) iodide
Show More Show Less
Hazard Category H301+H331-H350
Hazard Statement GHS H Statement
H301-H331
Toxic if swallowed.
Toxic if inhaled.
Precautionary Statement P201-P202-P261-P264b-P270-P271-P281-P301+P310-P304+P340-P308+P313-P311-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ARSENIC COMPOUNDS, SOLID, N.O.S.
EINECSNumber 232-068-4
RTECSNumber CG1950000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only