accessibility menu, dialog, popup

UserName

CAS RN 7784-45-4

IUPAC Name: triiodoarsane
Synonyms: arsenic triiodide
Molecular Weight (g/mol): 455.64
Molecular Formula: AsI3
InChi Key: IKIBSPLDJGAHPX-UHFFFAOYSA-N
SMILES: I[As](I)I
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)