Learn More
Arsenic(III) iodide, 98%
CAS: 7784-45-4 | AsI3 | 455.635 g/mol
$293.04 - $963.86
Chemical Identifiers
| CAS | 7784-45-4 |
|---|---|
| Molecular Formula | AsI3 |
| Molecular Weight (g/mol) | 455.635 |
| MDL Number | MFCD00014170 |
| InChI Key | IKIBSPLDJGAHPX-UHFFFAOYSA-N |
| Synonym | arsenous triiodide, arsenic iii iodide, arsenic iodide, arsenic triiodide, triiodoarsine, arsenous iodide, hsdb 420, arsen iii-jodid |
| PubChem CID | 24575 |
| IUPAC Name | triiodoarsane |
| SMILES | [As](I)(I)I |
Description
Arsenic(III) iodide is mainly used to treat rheumatism, arthritis, malaria, trypanosome infections, tuberculosis, and diabetes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 7784-45-4 | |
| 455.635 | |
| IKIBSPLDJGAHPX-UHFFFAOYSA-N | |
| 24575 | |
| [As](I)(I)I |
| AsI3 | |
| MFCD00014170 | |
| arsenous triiodide, arsenic iii iodide, arsenic iodide, arsenic triiodide, triiodoarsine, arsenous iodide, hsdb 420, arsen iii-jodid | |
| triiodoarsane |
Specifications
| 424°C | |
| 7784-45-4 | |
| 10 g | |
| MFCD00014170 | |
| 14,803 | |
| Soluble in alcohol,ether,Carbon disulfide,water(6g/100 m). | |
| [As](I)(I)I | |
| 455.635 | |
| 455.64 | |
| Pungent | |
| 4.39 g/mL |
| 140°C | |
| Pieces | |
| AsI3 | |
| UN1557 | |
| arsenous triiodide, arsenic iii iodide, arsenic iodide, arsenic triiodide, triiodoarsine, arsenous iodide, hsdb 420, arsen iii-jodid | |
| IKIBSPLDJGAHPX-UHFFFAOYSA-N | |
| triiodoarsane | |
| 24575 | |
| 98% | |
| Moisture sensitive | |
| Arsenic (III) iodide |
Safety and Handling
GHS H Statement
H301-H331
Toxic if swallowed.
Toxic if inhaled.
P201-P202-P261-P264b-P270-P271-P281-P301+P310-P304+P340-P308+P313-P311-P330-P501c
H301+H331-H350
DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ARSENIC COMPOUNDS, SOLID, N.O.S.
EINECSNumber : 232-068-4
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only