missing translation for 'onlineSavingsMsg'
Learn More

Arsenazo III

Catalog No. p-7042581
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6303106 5 g
AAJ6303114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog No. AAJ6303106 Supplier Thermo Scientific Chemicals Supplier No. J6303106
Only null left
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.

Arsenazo III (3,6-bis((E)-(2-Arsonophenyl)diazenyl)-4,5-dihydroxynaphthalene-2,7-disulfonic acid, CAS # 1668-00-4) is a metallochrome indicator that is used for detecting calcium levels in plasma and serum. | CAS: 1668-00-4 | C22H18As2N4O14S2 | 776.363 g/mol

Suitable for determination of micromolar amounts of calcium

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Arsenazo III is highly specific for calcium and produces absorbance peaks in the 600-660 nm range
• It reacts in slightly acidic conditions, and is soluble in alcohol and semi-soluble in water

Application

• Arsenazo III can detect calcium in permeabilized cells; it is also capable of detecting uranium and thorium
• Arsenazo III has been used in vitro to measure intracellular Ca2+ levels in live neurons alongside patch clamp methods

Chemical Identifiers

CAS 1668-00-4
Molecular Formula C22H18As2N4O14S2
Molecular Weight (g/mol) 776.363
MDL Number MFCD00036695
InChI Key TVMZRHVOFZTNET-RIRMOVKSSA-N
Synonym arsenazo iii, arsenazo iii sodium salt, 2,2'-1, 8-dihydroxy-3,6-disulfo-2, 7-naphthalene-bis azo dibenzenearsonic acid, 2,7-bis 2-arsonophenylazo-1, 8-dihydroxy-3, 6-naphthalene disulfonic acid, 2,7-bis 2-arsonophenylazo-1,8-dihydroxynaphthalene-3,6-disulfonic acid, 2,7-bis 2-arsonophenylazo chromotropic acid, 2,2'-1,8-dihydroxy-3,6-disulfonaphthylene-2,7-bisazo bisbenzenearsonic acid, 3,6-bis 2-2-arsonophenyl diazen-1-yl-4,5-dihydroxynaphthalene-2,7-disulfonic acid, 3,6-bis 2-2-arsonophenyl diazenyl-4,5-dihydroxy-2,7-naphthalenedisulfonic acid, 3,6-bis 2-2-arsonophenyl hydrazin-1-ylidene-4,5-dioxonaphthalene-2,7-disulfonic acid
PubChem CID 9810878
IUPAC Name (3Z,6E)-3,6-bis[(2-arsonophenyl)hydrazinylidene]-4,5-dioxonaphthalene-2,7-disulfonic acid
SMILES C1=CC=C(C(=C1)NN=C2C(=CC3=C(C2=O)C(=O)C(=NNC4=CC=CC=C4[As](=O)(O)O)C(=C3)S(=O)(=O)O)S(=O)(=O)O)[As](=O)(O)O

Specifications

Color Brown to Purple
Quantity 5 g
UN Number UN3465
Beilstein 5717957
Formula Weight 776.37
Physical Form Powder
Chemical Name or Material Arsenazo III
Hazard Category H301+H331
Hazard Statement GHS H Statement
H301-H331
Toxic if swallowed.
Toxic if inhaled.
Precautionary Statement P261-P264b-P270-P271-P301+P310-P304+P340-P311-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOARSENIC COMPOUND, SOLID, N.O.S.
EINECSNumber 216-788-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only