missing translation for 'onlineSavingsMsg'
Learn More

Arsenazo III Indicator, Certified, LabChem™

Supplier:  LabChem LC114707

Catalog No. LC114707


Only null left

Description

Description

Specifications

Specifications

Arsenazo III Indicator
99.8,0.2
C22H18As2N4O14S2
Passes Test
C1=CC=C(C(=C1)NN=C2C(=CC3=C(C2=O)C(=O)C(=NNC4=CC=CC=C4[As](=O)(O)O)C(=C3)S(=O)(=O)O)S(=O)(=O)O)[As](=O)(O)O
776.363
776.37
Poly Bottle
Arsenic and its oxides; Carbon monoxide; Carbon dioxide; Nitrogen oxides; Sulfur compounds
Purple/Red
Liquid
1668-00-4 , 7732-18-5
C22H18As2N4O14S2
Soluble in water
TVMZRHVOFZTNET-RIRMOVKSSA-N
(3Z,6E)-3,6-bis[(2-arsonophenyl)hydrazinylidene]-4,5-dioxonaphthalene-2,7-disulfonic acid
9810878
Certified
1g/mL
1g/mL
125 mL

Chemical Identifiers

1668-00-4
776.363
9810878
C1=CC=C(C(=C1)NN=C2C(=CC3=C(C2=O)C(=O)C(=NNC4=CC=CC=C4[As](=O)(O)O)C(=C3)S(=O)(=O)O)S(=O)(=O)O)[As](=O)(O)O
C22H18As2N4O14S2
TVMZRHVOFZTNET-RIRMOVKSSA-N
(3Z,6E)-3,6-bis[(2-arsonophenyl)hydrazinylidene]-4,5-dioxonaphthalene-2,7-disulfonic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Solution is not hazardous.

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions