missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Arsenazo Iii, Honeywell Fluka™
for spectrophotometric det. of Th, Zr, U, Cd, Zn, Ca
$1936.66 - $1936.66
Chemical Identifiers
| CAS | 1668-00-4 |
|---|---|
| Molecular Formula | C22H18As2N4O14S2 |
| Molecular Weight (g/mol) | 776.363 |
| MDL Number | MFCD00036695 |
| InChI Key | TVMZRHVOFZTNET-RIRMOVKSSA-N |
| PubChem CID | 9810878 |
| IUPAC Name | (3Z,6E)-3,6-bis[(2-arsonophenyl)hydrazinylidene]-4,5-dioxonaphthalene-2,7-disulfonic acid |
| SMILES | C1=CC=C(C(=C1)NN=C2C(=CC3=C(C2=O)C(=O)C(=NNC4=CC=CC=C4[As](=O)(O)O)C(=C3)S(=O)(=O)O)S(=O)(=O)O)[As](=O)(O)O |
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
| 1668-00-4 | |
| 776.363 | |
| TVMZRHVOFZTNET-RIRMOVKSSA-N | |
| (3Z,6E)-3,6-bis[(2-arsonophenyl)hydrazinylidene]-4,5-dioxonaphthalene-2,7-disulfonic acid |
| C22H18As2N4O14S2 | |
| MFCD00036695 | |
| 9810878 | |
| C1=CC=C(C(=C1)NN=C2C(=CC3=C(C2=O)C(=O)C(=NNC4=CC=CC=C4[As](=O)(O)O)C(=C3)S(=O)(=O)O)S(=O)(=O)O)[As](=O)(O)O |
Specifications
| 1668-00-4 | |
| (HO)2C10H2(SO3H)2(N=NC6H4AsO3H2)2 | |
| 3465 | |
| TVMZRHVOFZTNET-RIRMOVKSSA-N | |
| (3Z,6E)-3,6-bis[(2-arsonophenyl)hydrazinylidene]-4,5-dioxonaphthalene-2,7-disulfonic acid | |
| 9810878 | |
| Glass Bottle | |
| Arsenazo Iii |
| C22H18As2N4O14S2 | |
| MFCD00036695 | |
| 5717957 | |
| C1=CC=C(C(=C1)NN=C2C(=CC3=C(C2=O)C(=O)C(=NNC4=CC=CC=C4[As](=O)(O)O)C(=C3)S(=O)(=O)O)S(=O)(=O)O)[As](=O)(O)O | |
| 776.363 | |
| 776.37g/mol | |
| 25 g |
Safety and Handling
P261-P273-P301 + P310-P311-P501
H301 + H331-H410
EINECSNumber : 216-788-6