missing translation for 'onlineSavingsMsg'
Learn More

Arsenazo Iii, Solstice

Catalog No. 6000357
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6000357 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 6000357 Supplier Solstice Supplier No. 1109025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

for spectrophotometric det. of Th, Zr, U, Cd, Zn, Ca

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 1668-00-4
Molecular Formula C22H18As2N4O14S2
Molecular Weight (g/mol) 776.363
MDL Number MFCD00036695
InChI Key TVMZRHVOFZTNET-RIRMOVKSSA-N
PubChem CID 9810878
IUPAC Name (3Z,6E)-3,6-bis[(2-arsonophenyl)hydrazinylidene]-4,5-dioxonaphthalene-2,7-disulfonic acid
SMILES C1=CC=C(C(=C1)NN=C2C(=CC3=C(C2=O)C(=O)C(=NNC4=CC=CC=C4[As](=O)(O)O)C(=C3)S(=O)(=O)O)S(=O)(=O)O)[As](=O)(O)O

Specifications

Packaging Glass Bottle
Linear Formula (HO)2C10H2(SO3H)2(N=NC6H4AsO3H2)2
UN Number 3465
Beilstein 5717957
Quantity 25 g
Formula Weight 776.37g/mol
Chemical Name or Material Arsenazo Iii
Hazard Statement P261-P273-P301 + P310-P311-P501
Precautionary Statement H301 + H331-H410
EINECSNumber 216-788-6