missing translation for 'onlineSavingsMsg'
Learn More

Arsenazo III, 0.2% (w/v), Ricca Chemical

Catalog No. R0794000120
Change view
Click to view available options
Quantity:
120 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R0794000120 120 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R0794000120 Supplier Ricca Chemical Company Supplier No. R0794000120A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1668-00-4 , 7732-18-5
Molecular Formula C22H18As2N4O14S2
Molecular Weight (g/mol) 776.363
InChI Key TVMZRHVOFZTNET-RIRMOVKSSA-N
PubChem CID 9810878
IUPAC Name (3Z,6E)-3,6-bis[(2-arsonophenyl)hydrazinylidene]-4,5-dioxonaphthalene-2,7-disulfonic acid
SMILES C1=CC=C(C(=C1)NN=C2C(=CC3=C(C2=O)C(=O)C(=NNC4=CC=CC=C4[As](=O)(O)O)C(=C3)S(=O)(=O)O)S(=O)(=O)O)[As](=O)(O)O

Specifications

CAS Min % 98.0, 0.2
CAS Max % 100.0, 0.2
Concentration or Composition (by Analyte or Components) 0.2% (w/v) Aqueous
Odor Odorless
Packaging Natural Poly Bottle
Color Pink
Quantity 120 mL
Chemical Name or Material Arsenazo III
ShelfLife 12 months