missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ergodyne™ Arsenal™ 5020 Standard Nylon Gear Duffel Bag
Arsenal™ 5020 Standard Gear Duffel Bag is designed to store, organize, and easily carry everything you need for the workday or weekend.
$69.95 - $106.33
Specifications
| Color | Red |
|---|---|
| Material | 600D Nylon |
| For Use With (Application) | Fire and Rescue, Construction, General Purpose |
| Quantity | 1 Ea. |
Description
- Durable material that withstands wear and tear
- Water-resistant backing helps to repel water and stains
- Heavy-duty webbing straps on top and side of duffel for extra reinforcement
- Removable and adjustable padded shoulder strap for easy transportation
- Heavy-duty zippers to secure gear inside the duffel
- Two accessory pockets for cell phones, headphones, pens, keys, and other small parts
Specifications
| Red | |
| 600D Nylon | |
| Fire and Rescue, Construction, General Purpose | |
| 1 Ea. |