missing translation for 'onlineSavingsMsg'
Learn More

Ergodyne™ Arsenal™ 5020 Standard Nylon Gear Duffel Bag

Catalog No. 19819722
Change view
Click to view available options
Volume (Metric):
54 L
112 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
19819722 54 L
19819731 112 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19819722 Supplier Ergodyne™ Supplier No. 13020

Arsenal™ 5020 Standard Gear Duffel Bag is designed to store, organize, and easily carry everything you need for the workday or weekend.

  • Durable material that withstands wear and tear
  • Water-resistant backing helps to repel water and stains
  • Heavy-duty webbing straps on top and side of duffel for extra reinforcement
  • Removable and adjustable padded shoulder strap for easy transportation
  • Heavy-duty zippers to secure gear inside the duffel
  • Two accessory pockets for cell phones, headphones, pens, keys, and other small parts

Specifications

Color Red
Material 600D Nylon
Dimensions (L x W x H) 21 x 12 x 12 in. (59 x 30 x 30 cm)
For Use With (Application) Fire and Rescue, Construction, General Purpose
Quantity 1 Ea.
Size Small
Volume (Metric) 54 L