missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Array Installation Kit for 3730 Genetic Analyzer
Array Install Kit with choice of 48 x 50cm or 48 x 36cm Capillary Length
Supplier: Thermo Fisher Scientific 4339249
Description
Kit choices
- 48 x 50cm Cap
- 48 x 36cm Cap
Order Info
1 Kit
Specifications
| 36 cm | |
| Array Installation Kit, 48 x 50cm Capillary Length, For 3730 Genetic Analyzer | |
| 3730 Genetic Analyzers | |
| Array Installation Kit |