missing translation for 'onlineSavingsMsg'
Learn More

Array Install Kits For 3730XL Genetic Analyzer

Catalog No. 4339251
Encompass
Change view
Click to view available options
Capillary Length:
36 cm
50 cm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capillary Length
4339251 36 cm
4339252 50 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 4339251 Supplier Thermo Fisher Scientific Supplier No. 4339251

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Contains 4 anode buffer containers

Array Install Kits with your choice of 96 x 50cm or 96 x 36cm Capillary Length

Kit choices

  • 96 x 50cm Cap
  • 96 x 36cm Cap

Order Info

1 Kit

Specifications

Capillary Length 36 cm
Description Array Install Kit, For 3730XL Genetic Analyzer
For Use With (Equipment) 3730XL Genetic Analyzers
Product Type Array Install Kit