missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ Array Calibration Ruler
Description
This is a spare part for the 3100 Genetic Analyzer.
Specifications
Specifications
| Content And Storage | Store at room temperature. |
| For Use With (Equipment) | 3100 Genetic Analyzer |
| For Use With (Application) | Sequencing |
| Product Type | Array Calibration Ruler |
| Quantity | 1 Ea. |
| Shipping Condition | Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.