missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ARPC4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5150086
Description
ARPC4 Polyclonal Antibody for Western Blot, ICC/IF, IHC
This gene encodes one of seven subunits of the human Arp2/3 protein complex. This complex controls actin polymerization in cells and has been conserved throughout eukaryotic evolution. This gene encodes the p20 subunit, which is necessary for actin nucleation and high-affinity binding to F-actin. Alternative splicing results in multiple transcript variants. Naturally occurring read-through transcription exists between this gene and the downstream tubulin tyrosine ligase-like family, member 3, which results in the production of a fusion protein.Chemical Identifiers
| ARPC4 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P59998, P59999 | |
| 10093, 297518, 68089 | |
| Recombinant Actin Related Protein 2/3 Complex Subunit 4 (ARPC4) (Ala79-Phe164). | |
| Antibody |
| 0.5 mg/mL | |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| ARPC4 | |
| 20kDa; 5330419I20Rik; actin related protein 2/3 complex subunit 4; actin related protein 2/3 complex subunit 4, 20kDa; actin related protein 2/3 complex, subunit 4; actin related protein 2/3 complex, subunit 4, 20kDa; actin-related protein 2/3 complex subunit 4; AI327076; ARC20; arp2/3 complex 20 kDa subunit; arp2/3 protein complex subunit p20; ARPC4; fa07b12; MGC13544; p20-ARC; wu:fa07b12; zgc:77932 | |
| ARPC4 | |
| Primary | |
| Protein A |
Specifications
| ARPC4 | |
| Polyclonal | |
| Unconjugated | |
| ARPC4 | |
| 20kDa; 5330419I20Rik; actin related protein 2/3 complex subunit 4; actin related protein 2/3 complex subunit 4, 20kDa; actin related protein 2/3 complex, subunit 4; actin related protein 2/3 complex, subunit 4, 20kDa; actin-related protein 2/3 complex subunit 4; AI327076; ARC20; arp2/3 complex 20 kDa subunit; arp2/3 protein complex subunit p20; ARPC4; fa07b12; MGC13544; p20-ARC; wu:fa07b12; zgc:77932 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 10093, 297518, 68089 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P59998, P59999 | |
| ARPC4 | |
| Recombinant Actin Related Protein 2/3 Complex Subunit 4 (ARPC4) (Ala79-Phe164). | |
| 200 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody |