missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Arp2 Recombinant Rabbit Monoclonal Antibody (23GB610)
GREENER_CHOICE

Catalog No. MA554052
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA554052 100 μL
MA554053 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA554052 Supplier Invitrogen™ Supplier No. MA554052
Only null left

Rabbit Recombinant Monoclonal Antibody

Arp2 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow

ARP2 (actin-related protein 2) is an ATP-binding component of the ARP2/3 complex - a multiprotein complex that mediates actin polymerization upon stimulation by the nucleation-promoting factor (NPF). The ARP2/3 complex mediates the formation of branched actin networks in the cytoplasm and provides the force for cell motility. The ARP2/3 complex also promotes actin polymerization in the nucleus, thereby regulating gene transcription and repair of damaged DNA.

Specifications

Antigen Arp2
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB610
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ACTR2
Gene Accession No. P61160, P61161, Q5M7U6
Gene Alias 4921510D23Rik; AA409782; Actin-like protein 14D; Actin-like protein 2; Actin-related protein 14 Da; Actin-related protein 14D; Actin-related protein 2; Actr14D; Actr2; Arp14D; ARP2; ARP2 actin related protein 2 homolog; ARP2 actin-related protein 2; ARP2 actin-related protein 2 homolog; ARP2 actin-related protein 2 homolog (yeast); Arp2-PA; Arp2-PB; Arp2-PC; CG9901; CG9901-PA; CG9901-PB; CG9901-PC; D6Ertd746e; Dmel\CG9901; Dmel_CG9901; I79_003843; LD18955p
Gene Symbols ACTR2
Host Species Rabbit
Immunogen A synthetic peptide of human Arp2 (350-394AA).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10097, 289820, 66713
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less