missing translation for 'onlineSavingsMsg'
Learn More

Aroclor 1262, Restek™

Catalog No. 06720008
Encompass
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06720008 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06720008 Supplier Chromatographic Supplier No. RK 32409

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 37324-23-5 , 110-54-3

Specifications

Packaging Ampule
Odor Mild
Vapor Density 2.97 (Air = 1)
Flash Point -8°C
Specific Gravity 0.672 g/cm3 at 15°C
Physical Form Liquid
Concentration 1000 μg/mL in hexane
UN Number UN1208
Melting Point -95°C
Type Single-component Standard
Quantity 1 mL
Chemical Name or Material Arochlor 1262
Show More Show Less
ShelfLife 6 months