missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aroclor 1248 (High Level), SPEX CertiPrep™
Description
Includes
Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
| Analyte | CAS No. | Concentration |
|---|---|---|
| Aroclor 1248 | 12672-29-6 | 1000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Specifications
| Packaging | 2mL amber prescored glass ampule |
| Concentration or Composition Notes | Certificate of Analysis reports target analytes to a ±2% accuracy and correct for raw material purity. |
| Methods | For US EPA Methods 505, 508, 625, 8082, 8270C |
| Technique | AA, ICP |
| For Use With (Equipment) | GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation |
| Type | Certified Reference Material |
| Solvent or Matrix | Hexane |
| Quantity | 1 mL |
| Concentration or Composition (by Analyte or Components) | 1000μg/mL |
| Chemical Name or Material | Aroclor 1248 (High Level) |
Safety and Handling
| Hazard Category | Harmful if swallowed. Suspected of causing cancer. |
| DOTInformation | Nonhazardous |
| Recommended Storage | Freezer |
Product intended for laboratory use only