missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| Aripiprazole | |
| 149°C | |
| MFCD00892072 | |
| aripiprazole, abilify, abilitat, abilify discmelt, discmelt, aripiprazol, opc 31, aripiprazole usan, aripiprazolum, aripirazole | |
| C1CC(=O)NC2=C1C=CC(=C2)OCCCCN3CCN(CC3)C4=C(C(=CC=C4)Cl)Cl | |
| 448.388 | |
| CHEBI:31236 | |
| ≥98.0% (HPLC,T) |
| 129722-12-9 | |
| C23H27Cl2N3O2 | |
| 5 g | |
| CEUORZQYGODEFX-UHFFFAOYSA-N | |
| 7-[4-[4-(2,3-dichlorophenyl)piperazin-1-yl]butoxy]-3,4-dihydro-1H-quinolin-2-one | |
| 60795 | |
| 448.39 | |
| Crystalline Powder |
Chemical Identifiers
| 129722-12-9 | |
| 448.388 | |
| CEUORZQYGODEFX-UHFFFAOYSA-N | |
| 60795 | |
| 7-[4-[4-(2,3-dichlorophenyl)piperazin-1-yl]butoxy]-3,4-dihydro-1H-quinolin-2-one |
| C23H27Cl2N3O2 | |
| MFCD00892072 | |
| aripiprazole, abilify, abilitat, abilify discmelt, discmelt, aripiprazol, opc 31, aripiprazole usan, aripiprazolum, aripirazole | |
| CHEBI:31236 | |
| C1CC(=O)NC2=C1C=CC(=C2)OCCCCN3CCN(CC3)C4=C(C(=CC=C4)Cl)Cl |
Safety and Handling
RTECSNumber : VC8275950
TSCA : No