missing translation for 'onlineSavingsMsg'
Learn More

Aripiprazole 98.0+%, TCI America™

Catalog No. p-7117942
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A24961G 1 g
A24965G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A24961G Supplier TCI America Supplier No. A24961G
Only null left

Chemical Identifiers

CAS 129722-12-9
Molecular Formula C23H27Cl2N3O2
Molecular Weight (g/mol) 448.388
MDL Number MFCD00892072
InChI Key CEUORZQYGODEFX-UHFFFAOYSA-N
Synonym aripiprazole, abilify, abilitat, abilify discmelt, discmelt, aripiprazol, opc 31, aripiprazole usan, aripiprazolum, aripirazole
PubChem CID 60795
ChEBI CHEBI:31236
IUPAC Name 7-[4-[4-(2,3-dichlorophenyl)piperazin-1-yl]butoxy]-3,4-dihydro-1H-quinolin-2-one
SMILES C1CC(=O)NC2=C1C=CC(=C2)OCCCCN3CCN(CC3)C4=C(C(=CC=C4)Cl)Cl

Specifications

Melting Point 149°C
Quantity 1 g
Formula Weight 448.39
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Aripiprazole
RTECSNumber VC8275950
TSCA No