missing translation for 'onlineSavingsMsg'
Learn More

Aripiprazole Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111012916
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012916 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012916 Supplier Merck Emd Millipore Supplier No. PHR1784500MG

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 129722-12-9
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C23H27 Cl2N3O2
Synonym Aripiprazole; 7-{4-[4-(2,3-Dichlorophenyl)-1-piperazinyl]butoxy}-3,4-dihydro-2(1 H)-quinolinone
Type Certified Reference Material
Quantity 500 mg
Formula Weight 448.39
Grade Certified Reference Material