missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARID1A Monoclonal Antibody (KAA140_16E7), Invitrogen™
Mouse Monoclonal Antibody
$1557.73 - $2745.56
Specifications
| Antigen | ARID1A |
|---|---|
| Clone | KAA140_16E7 |
| Concentration | 1 mg/mL |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
Description
ARID1A Monoclonal antibody specifically detects ARID1A in Human samples. It is validated for Immunoprecipitation, Western BlotSpecifications
| ARID1A | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| 1110030E03Rik; ARID domain-containing protein 1A; ARID1A; AT rich interactive domain 1A (Swi1 like); AT rich interactive domain 1A (SWI-like); AT-rich interaction domain 1A; AT-rich interactive domain-containing protein 1A; B120; BAF250; Baf250a; BM029; brain protein 120; BRG1-associated factor 250; BRG1-associated factor 250a; C1orf4; chromatin remodeling factor p250; CSS2; ELD; h BM029; hELD; hOSA1; MRD14; Osa homolog 1; Osa1; OSA1 nuclear protein; P270; Smarcf1; SWI/SNF complex protein p270; SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily f, member 1; SWI/SNF-related, matrix-associated, actin-dependent regulator of chromatin subfamily F member 1; Swi1 like; SWI-like protein; SWI-SNF complex protein p270 | |
| Recombinant human ARID1A (proprietary range information). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| ARID1A |
| KAA140_16E7 | |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| O14497 | |
| 8289 | |
| IgG1 | |
| Protein A | |
| Antibody |