missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ARHGEF7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA575963
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
The serine/threonine kinase, p21 activated kinase (PAK), is a downstream effector of the small GTPases Cdc42 and Rac. PAK associates with Nck, the p85 and p110 subunits of PI3-kinase and PIX (PAK-interacting exchange factor) in a focal complex. The binding of PIX is necessary for the localization and activation of PAK in the Cdc42 to Rac signaling pathway, and this binding occurs through the high affinity of the N-terminal SH3 domain of PIX for a conserved proline rich PAK sequence. PIX exists as two isoforms, alpha and beta, and both are highly expressed in heart, muscle and thymus tissues of human and rat. alphaPIX is phosphorylated via PDGF and EphB2 receptor signaling pathways or through association with PI3-kinase. The alphaPIX isoform predominantly acts as a guanine nucleotide exchange factor (GEF) on Rac, which may mediate lamellipodia formation.Chemical Identifiers
| ARHGEF7 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Human, Mouse, Monkey | |
| Arhgef7 | |
| ARHGEF7; betaPix; Beta-Pix; betaPix-b; betaPix-c; Cool; COOL1; cool-1; KIAA0142; mKIAA0142; Nbla10314; P50; P50BP; p85; p85Cool1; p85SPR; PAK3; Pak3 binding protein; Pak3bp; PAK-interacting exchange factor beta; PIX; Pixb; PIXbeta; Rho guanine nucleotide exchange factor (GEF) 7; Rho guanine nucleotide exchange factor (GEF7); Rho guanine nucleotide exchange factor 7; SH3 domain-containing proline-rich protein | |
| Arhgef7 | |
| Primary | |
| Antigen Affinity Chromatography |
| 1 mg/mL | |
| Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q14155, Q9ES28 | |
| 54126, 8874 | |
| Recombinant full length Human ARHGEF7. | |
| Antibody |
Specifications
| ARHGEF7 | |
| Polyclonal | |
| Unconjugated | |
| Arhgef7 | |
| ARHGEF7; betaPix; Beta-Pix; betaPix-b; betaPix-c; Cool; COOL1; cool-1; KIAA0142; mKIAA0142; Nbla10314; P50; P50BP; p85; p85Cool1; p85SPR; PAK3; Pak3 binding protein; Pak3bp; PAK-interacting exchange factor beta; PIX; Pixb; PIXbeta; Rho guanine nucleotide exchange factor (GEF) 7; Rho guanine nucleotide exchange factor (GEF7); Rho guanine nucleotide exchange factor 7; SH3 domain-containing proline-rich protein | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 54126, 8874 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q14155, Q9ES28 | |
| Arhgef7 | |
| Recombinant full length Human ARHGEF7. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Monkey | |
| Antibody | |
| IgG |