missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ARHGEF18 Monoclonal Antibody (OTI6D10)
GREENER_CHOICE

Catalog No. PIMA527295
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA527295 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA527295 Supplier Invitrogen™ Supplier No. MA527295
Only null left

Mouse Monoclonal Antibody

ARHGEF18 Monoclonal Antibody for Western Blot, IHC (P)

Rho GTPases are GTP binding proteins that regulate a wide spectrum of cellular functions. These cellular processes include cytoskeletal rearrangements, gene transcription, cell growth and motility. Activation of Rho GTPases is under the direct control of guanine nucleotide exchange factors (GEFs). The protein encoded by this gene is a guanine nucleotide exchange factor and belongs to the Rho GTPase GFE family. Family members share a common feature, a Dbl (DH) homology domain followed by a pleckstrin (PH) homology domain. Alternatively spliced transcript variants encoding different isoforms have been identified.

Specifications

Antigen ARHGEF18
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI6D10
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Arhgef18
Gene Accession No. Q6ZSZ5
Gene Alias 114 kDa Rho-specific guanine nucleotide exchange factor; AI467246; Arhgef18; D030053O22Rik; KIAA0521; p114RhoGEF; P114-RhoGEF; p114-Rho-GEF; rho guanine nucleotide exchange factor 18; rho/rac guanine nucleotide exchange factor (GEF) 18; Rho/Rac guanine nucleotide exchange factor 18; Rho-specific guanine nucleotide exchange factor p114; SA-RhoGEF; Septin-associated RhoGEF
Gene Symbols Arhgef18
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 118-215 of ARHGEF18 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23370
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less