missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARHGAP10 Polyclonal Antibody

ARHGAP10 Polyclonal Antibody
Supplier: Thermo Fisher Scientific PA524188
Description
ARHGAP10 Polyclonal Antibody for Western Blot, IHC (P), Flow
Specifications
| ARHGAP10 | |
| Polyclonal | |
| Unconjugated | |
| ARHGAP10 | |
| A930033B01Rik; Arhgap10; GRAF2; Graf-related protein 2; GTPase regulator associated with focal adhesion kinase 2; PH and SH3 domain-containing rhoGAP protein; PSGAP; PS-GAP; PSGAP-m; PSGAP-s; Rho GTPase activating protein 10; rho GTPase-activating protein 10; Rho-type GTPase-activating protein 10 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 79658 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| A1A4S6 | |
| ARHGAP10 | |
| KLH conjugated synthetic peptide between 237-263 amino acids from the central region of human ARHGAP10. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |