Learn More
Synthware™ Argon/Nitrogen Gas Manifold with PTFE Stopcocks
GREENER_CHOICE
Catalog No. p-7179984
Change view
Click to view available options
Type:
Manifold
Nitrogen/Argon
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
31501792 Nitrogen/Argon
31501793 Manifold
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 31501792 Supplier Synthware™ Supplier No. M531042
Only null left
With 14/20 outer joint at the center to fit a rubber septum for keeping syringes under an inert environment cutting down on the number used.
Specifications
Connection Type 4-port; 2 mm
Type Nitrogen/Argon
For Use With (Application) Organic Synthesis
Includes PTFE Stopcocks
Material Borosilicate Glass