missing translation for 'onlineSavingsMsg'
Learn More

ARG1 293T Cell Overexpression Lysate 2 (Denatured), Abnova

Catalog No. 89000260
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89000260 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89000260 Supplier Abnova Corporation Supplier No. H00000383T02

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Western Blotting

Specifications

For Use With (Application) Western Blot
Cell Line 293T
Description ARG1 293T Cell Transient Overexpression Lysate(Denatured)
Gene Accession No. NM_000045.2
Molecular Weight (g/mol) 35.53
pH Range 6.8
Plasmid pCMV-ARG1 full-length
Quality Control Testing Transient overexpression cell Lysate was tested with Anti-ARG1 antibody (H00000383-B02) by western blotting
Quantity 100 μL
Storage Buffer 1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Host Species Human
Content And Storage Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Show More Show Less