Learn More
Invitrogen™ ARES™ Alexa Fluor™ 647 DNA Labeling Kit
Description
ARES™ Alexa Fluor™ Labeling Kit Specifications:
More Uniform Labeling With ARES™ Labeling Kits
ARES™ Alexa Fluor™ Labeling Kits employ a two-step labeling technology—nick translation to enzymatically incorporate an amine-modified nucleotide (aminoallyl dUTP) followed by chemical labeling with Alexa Fluor™ dyes. This method achieves uniformity and consistency of labeling that is difficult to obtain with conventional enzymatic incorporation of labeled nucleotides.
We also offer this two-step labeling technology in our FISH Tag™ DNA and FISH Tag™ RNA Kits, which provide a complete workflow solution for FISH applications, including all of the reagents for probe synthesis, labeling, purification and an anti-fade reagent to help protect the signal during fluorescence microscopy.
More Options for Nucleic Acid Labeling
To review various options for nucleic acid labeling (including DNA and RNA FISH), review Labeling Oligonucleotides and Nucleic Acids—Section 8.2 in the Molecular Probes™ Handbook.
For Research Use Only. Not intended for human or animal therapeutic or diagnostic use.
Specifications
Specifications
| Content And Storage | Store in freezer (-5 to -30°C) and protect from light. |
| Detection Method | Fluorescence |
| Format | Kit |
| Label or Dye | Alexa Fluor 647 |
| Final Product Type | Probes (Labeled DNA), cDNA (Labeled) |
| Labeling Target | DNA (General), cDNA |
| Includes Label or Dye | Yes |
| Labeling Method | Indirect Labeling |
| Product Line | ARES, Alexa Fluor |
| Product Type | DNA Labeling Kit |
| Show More |
Frequently Asked Questions (FAQs)
Different preparations of RNA will certainly give different results. Most of the time, the mRNA is significantly degraded. The enzymatic incorporation of aminoallyl-dUTP (AA-dUTP) should not differ from reaction to reaction. If there are differences, it has to be due to the RNA or the method. AA-dUTP incorporation is no different than that of a dye-nucleotide conjugate, and should be more efficient and uniform. Here are a couple of suggestions:
1) cDNA may have been lost prior to labeling. Add 1 µL of glycogen (molecular biology grade), containing 10-20 µg, to the cDNA before precipitating it with ethanol.
2) Make sure to add sodium acetate as the salt and not ammonium acetate. After pelleting the cDNA, resuspend it in 5 µL water.
3) If generating long cDNAs, it will help to heat-denature the sample. Heat it at 95°C for 5 minutes and then put it on ice for a few minutes. Then centrifuge it for a few minutes just prior to the labeling reaction.
4) You want the tube to be at room temperature for the labeling reaction. Add the 3 µL of buffer and mix it in. Then add the dye and vortex it vigorously for at least 15 seconds.
Unfortunately, Alexa Fluor 633 does not label nucleic acids well because of the dye's chemical structure. Furthermore, DNA probes labeled with Alexa Fluor 633 do not form stable hybrids in nucleic acid hybridization assays.
An ARES-labeled oligonucleotide should survive at 95°C for 5 minutes.
Long-term storage for the ARES-labeled probes can be done in just about any kind of buffer, TE, formamide, hybridization buffer, or ethanol. We suggest using your normal storage conditions as long as you protect the probes from light. ARES conjugates are very stable.
At the same dye-to-base ratio, Alexa Fluor dyes exhibit higher intensity and reduced self-quenching at higher labeling ratios.
For Research Use Only. Not for use in diagnostic procedures.