Learn More
Heathrow Scientific™ Arctic Squares™ Cryogenic Tube Storage Box
Safely store cryogenic vials in this Arctic Squares™ Cryogenic Tube Storage Box.
Specifications
| Material | Polycarbonate |
|---|---|
| Shape | Square |
| Type | Cryo Box |
| Holds | 2.0 mL Cryogenic Vials |
| Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity & Availability | |||||
17100462
![]() |
Heathrow Scientific™
HS23300D |
100 Cryogenic Vials |
N/A
|
|
|||||
17100446
![]() |
Heathrow Scientific™
HS23325A |
25 Cryogenic Vials |
N/A
|
|
|||||
17100427
![]() |
Heathrow Scientific™
HS23381B |
81 Cryogenic Vials |
N/A
|
|
|||||
Description
- Our polycarbonate boxes are designed with unique features to help orient, identify and access vials with ease
- Alphanumeric labeling on lid or base (depending on the unit) for quick location and retrieval of tubes
- Liquid nitrogen vent and drainage holes
- Magnet on top of forceps enables easy storing on metal surfaces
- Stackable, saves valuable space in the lab
Specifications
| Polycarbonate | |
| Square | |
| Cryo Box | |
| 2.0 mL Cryogenic Vials |