missing translation for 'onlineSavingsMsg'
Learn More

National Safety Apparel™ Enespro™ 25 kA ArcGuard™ Supression Blanket Kit, 4 ft x 5 ft

Ideal solution for jobs in confined and enclosed spaces where arc flash is a hazard. It is available on its own, or in a convenient kit package which includes the blanket, straps, and carrying bags

$3,856.17 - $4,862.59

Specifications

Product Type Fire Blanket
Color Black
Material Multi-layer Material with Reinforced Holes
Disposable Reusable
For Use With (Application) Electrical Contractors
View More Specs

Products 2
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity & Availability  
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity & Availability  
17001749
View Documents Promotion Details
National Safety Apparel™
K25KIT4F5F
48 x 60 in.
Each for $3,856.17
Only null left
 
17001750
View Documents Promotion Details
National Safety Apparel™
K25KIT4F8F
48 x 96 in.
Each for $4,862.59
Only null left
 
Description

Description

  • 12 attachment points
  • Multi-layer material with reinforced holes for straps
  • Provides optimal barrier in enclosed and confined spaces
  • Breakopen Threshold Performance (BTP) = 362 kA cycles
  • Maximum Arc current Imax = 25 kA
  • Kit includes 4 ft x 5 ft arc suppression blanket, straps and gear bag
  • Made in the USA
Specifications

Specifications

Fire Blanket
Multi-layer Material with Reinforced Holes
Electrical Contractors
Black
Reusable
Arc Supression Blanket, Straps, and Gear Bag
Videos
SDS
Documents

Documents