missing translation for 'onlineSavingsMsg'
Learn More

National Safety Apparel™ Enespro™ 25 kA ArcGuard™ Supression Blanket Kit, 4 ft x 5 ft

Catalog No. 17001749
Change view
Click to view available options
Dimensions (L x W):
48 x 60 in.
48 x 96 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
17001749 48 x 60 in.
17001750 48 x 96 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17001749 Supplier National Safety Apparel™ Supplier No. K25KIT4F5F

Ideal solution for jobs in confined and enclosed spaces where arc flash is a hazard. It is available on its own, or in a convenient kit package which includes the blanket, straps, and carrying bags

  • 12 attachment points
  • Multi-layer material with reinforced holes for straps
  • Provides optimal barrier in enclosed and confined spaces
  • Breakopen Threshold Performance (BTP) = 362 kA cycles
  • Maximum Arc current Imax = 25 kA
  • Kit includes 4 ft x 5 ft arc suppression blanket, straps and gear bag
  • Made in the USA

Specifications

Product Type Fire Blanket
Color Black
Material Multi-layer Material with Reinforced Holes
Certifications/Compliance NFPA 70 E, ASTM F2700, CSA Z462, OSHA 1910.269, ASTM F1506
Dimensions (L x W) 48 x 60 in.
Disposable Reusable
For Use With (Application) Electrical Contractors
Includes Arc Supression Blanket, Straps, and Gear Bag