missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Arc Flash Labels - Self-sticking Polyester

Catalog No. 19104670
Change view
Click to view available options
Display Text:
DANGER Arc Flash And Shock Hazard Appropriate PPE Required
DANGER Arc Flash And Shock Hazard Appropriate PPE Required (with picto)
WARNING Arc Flash And Shock Hazard Appropriate PPE Required
WARNING Arc Flash And Shock Hazard Appropriate PPE Required (with picto)
Quantity:
100/Roll
5 Pk.
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Display Text Quantity
19104670 WARNING Arc Flash And Shock Hazard Appropriate PPE Required (with picto) 5 Pk.
19104672 WARNING Arc Flash And Shock Hazard Appropriate PPE Required 5 Pk.
19104673 DANGER Arc Flash And Shock Hazard Appropriate PPE Required 5 Pk.
19104676 WARNING Arc Flash And Shock Hazard Appropriate PPE Required (with picto) 100/Roll
19104671 DANGER Arc Flash And Shock Hazard Appropriate PPE Required (with picto) 5 Pk.
19104678 WARNING Arc Flash And Shock Hazard Appropriate PPE Required 100/Roll
19104677 DANGER Arc Flash And Shock Hazard Appropriate PPE Required (with picto) 100/Roll
19104679 DANGER Arc Flash And Shock Hazard Appropriate PPE Required 100/Roll
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 19104670 Supplier Brady™ Supplier No. 101950

Self-sticking Polyester (B-302) is a flexible polymer plastic over-laminated with a clear plastic coating to protect graphics against abrasion, sunlight, chemicals.

  • Weathers up to 8 years outdoors, even longer indoors, and withstands wide temperature variations (up to 230°F).
  • Flexible material conforms to most surfaces while protected graphics hold up to harsh environments, abrasion, and repeated spills and cleanings.
  • Permanent, pressure sensitive adhesive forms a long-lasting bond to even the most difficult surfaces and shapes.
  • Cold temperature adhesive allows the signage to be applied at 0°F (-18°C).

Specifications

Autoclavable Not Autoclavable
Certifications/Compliance NEC, NFPA 70E, OSHA 1910.331-335
Description Self-adhesive Label
Adhesive Type Permanent Acrylic
Color Black, Orange on White
Finish Gloss
Format Label
Material Polyester
Printable Sides Single-side Printable
Product Type Label
Sterility Non-sterile
Display Text WARNING Arc Flash And Shock Hazard Appropriate PPE Required (with picto)
Includes Pack of 5 Labels
Languages English
Quantity 5 Pk.
Shape Rectangle
Size (English) 5 x 3.5 in. (W x H)
Thickness (English) 6 mil
Thickness (Metric) 0.152 mm
Dimensions (L x W) 3.5 x 5 in.
Width (English) 5 in.
Width (Metric) 127.00 mm
Height (English) 3.5 in.
Height (Metric) 88.90 mm
Show More Show Less