missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Arachidonyl-L-carnitine, ≥95.0% (LC/MS), MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 36816-11-2 |
| Linear Formula | C27H45 NO4 |
| Synonym | (2 R)-3-Carboxy-N,N,N-trimethyl-2-[[(5 Z,8 Z,11 Z,14 Z)-1-oxo-5,8,11,14-eicosatetraen-1-yl]oxy]-1-propanaminium inner salt; Arachidonoylcarnitine |
| Type | Analytical Standard |
| Quantity | 10 mg |
| Formula Weight | 447.65 |
| Percent Purity | ≥95.0% (LC/MS) |
| Grade | Analytical Standard |