missing translation for 'onlineSavingsMsg'
Learn More

Arachidonic acid, ≥97.0% (GC), MilliporeSigma™ Supelco™

Catalog No. 111014369
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014369 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014369 Supplier Merck Emd Millipore Supplier No. 2340150MG
Only null left
Food and Beverages

Specifications

CAS 506-32-1
Density 0.922 g/mL (at 25°C)
Flash Point 113°C - Closed Cup
Physical Form Liquid
Linear Formula CH3(CH2)4(CH=CHCH2)4 CH2 CH2 CO2 H
MDL Number MFCD00004417
Melting Point -49°C
Boiling Point 169°C to 171°C (0.15 mmHg)
Synonym cis,cis,cis,cis-5,8,11,14-Eicosatetraenoic acid; Eicosa-5 Z,8 Z,11 Z,14 Z-tetraenoic acid; Immunocytophyte
Type Analytical Standard
Quantity 50 mg
Formula Weight 304.47
Percent Purity ≥97.0% (GC)
Grade Analytical Standard
Show More Show Less