missing translation for 'onlineSavingsMsg'
Learn More

OHAUS™ Aquasearcher™ pH Meter AB23PH

The Aquasearcher™ pH Meter AB23PH is a simple to use benchtop meter for measuring pH and oxidation-reduction potential (ORP). An intuitive design and auto-buffer recognition make measuring pH straightforward and easy.

$1,081.59 - $1,230.59

Specifications

Accuracy (mV) ±1
Accuracy (pH) ±0.01
Accuracy (Temperature) ±0.5°C
Data Management Details Datalog for Measurements and Calibration
Measurement Modes Auto-stop, Continuous
View More Specs

Products 2
Catalog Number Mfr. No. Type Price Quantity & Availability  
Catalog Number Mfr. No. Type Price Quantity & Availability  
01922375
View Documents Promotion Details
OHAUS™
30589820
pH Meter
Each for $1,081.59
Only null left
 
01922374
View Documents Promotion Details
OHAUS™
30589821
Meter with pH Kit
Each for $1,230.59
Only null left
 
Description

Description

  • Equipped with a large 5 in. LCD display and six instructional menu buttons
  • Auto buffer recognition and 3-point calibration capability ensures a reliable calibration result
  • Features a 99 item memory for measurements and last calibration
  • Choose between just the AB23PH meter with electrode holder and a full kit including AB23PH meter, ST320 pH electrode, and four pH Buffer Mini Kits (50 mL)
Specifications

Specifications

±1
±0.5°C
Auto-stop, Continuous
0 to 14
±1999
0.0°C to 100.0°C
Yes
Automatic Data Logging
±0.01
Datalog for Measurements and Calibration
Linear
0.01
Up to 3
LCD
99 Sets
Videos
SDS
Documents

Documents