missing translation for 'onlineSavingsMsg'
Learn More

OHAUS™ Aquasearcher™ pH Meter AB23PH

Catalog No. 01922375
Change view
Click to view available options
Type:
Meter with pH Kit
pH Meter
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
01922375 pH Meter
01922374 Meter with pH Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01922375 Supplier OHAUS™ Supplier No. 30589820

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The Aquasearcher™ pH Meter AB23PH is a simple to use benchtop meter for measuring pH and oxidation-reduction potential (ORP). An intuitive design and auto-buffer recognition make measuring pH straightforward and easy.

  • Equipped with a large 5 in. LCD display and six instructional menu buttons
  • Auto buffer recognition and 3-point calibration capability ensures a reliable calibration result
  • Features a 99 item memory for measurements and last calibration
  • Choose between just the AB23PH meter with electrode holder and a full kit including AB23PH meter, ST320 pH electrode, and four pH Buffer Mini Kits (50 mL)

Specifications

Data Management Details Datalog for Measurements and Calibration
Accuracy (pH) ±0.01
Accuracy (mV) ±1
Accuracy (Temperature) ±0.5°C
Includes AB23PH Meter, Electrode Holder
Measurement Modes Auto-stop, Continuous
Calibration Linear
Range (pH) 0 to 14
Resolution (pH) 0.01
Range (mV) ±1999
Calibration Points Up to 3
Temperature Range (Metric) 0.0°C to 100.0°C
Display Type LCD
Backlight Option Yes
Data Points 99 Sets
Log Function Type Automatic Data Logging
Type pH Meter
Show More Show Less