missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OHAUS™ Aquasearcher™ Multi-Parameter Analyzer AB33M1
The Aquasearcher™ Multi-Parameter Analyzer AB33M1 is easy-to-use and accurate, capable of measuring up to seven parameters including pH, ORP, conductivity, TDS, salinity, resistivity, and temperature.
$2,168.70 - $2,552.24
Specifications
| Outputs | RS232, USB Device (Included) |
|---|---|
| Channels | pH, Conductivity |
| Display Type | LCD |
| IP Rating | None |
| Measurement Modes | Auto-stop, Continuous |
Description
- Features two independent measuring channels which can work simultaneously
- Equipped with a large 6.5 in. LCD display and a touch keypad
- Features a 1000 item memory for measurements and calibration trail
- Provides connectivity and data output capabilities with RS232 and USB interface
- i-Steward monitors the condition of electrodes to ensure repeatable and consistent measurement accuracy
- Intuitive user interface makes changing the parameters, executing setup, and performing calibration as simple as using a smartphone
- Choose between just the AB33M1 meter with electrode holder and a full kit including AB33M1 meter, ST310 electrode, STCON3 probe, electrode holder, 4 pH buffer mini kits (50 mL), standard conductivity solution kits (1413 μS/cm, 50 mL)
Specifications
| RS232, USB Device (Included) | |
| LCD | |
| Auto-stop, Continuous | |
| -2000 to 2000 mV | |
| 2 to 100 MΩ-cm | |
| 0.1 mg/L to 199.9 g/L | |
| 0.1/0.01 pH |
| pH, Conductivity | |
| None | |
| 0.01 μS/cm to 500 mS/cm | |
| -2 to 20 | |
| 0 to 100 psu | |
| 0.01 pH | |
| -5°C to +110°C |