missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aquaporin 10 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Fisher Scientific PA564252
Description
Immunogen sequence: LVALHHPEGP EPAQDLVSAQ HKASELETPA SAQMLECKL Highest antigen sequence identity to the following orthologs: Mouse - 34%, Rat - 37%.
Aquaporin 10 Polyclonal antibody specifically detects Aquaporin 10 in Human samples. It is validated for Immunohistochemistry.Chemical Identifiers
| Aquaporin 10 | |
| Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q96PS8 | |
| 89872 | |
| IgG | |
| Antigen affinity chromatography | |
| Antibody |
| 0.05 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| AQP10; AQP-10; AQPA_HUMAN; Aquaglyceroporin-10; aquaporin 10; aquaporin-10; Small intestine aquaporin | |
| AQP10 | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| AQP10 |
Specifications
| Aquaporin 10 | |
| Polyclonal | |
| Unconjugated | |
| AQP10 | |
| AQP10; AQP-10; AQPA_HUMAN; Aquaglyceroporin-10; aquaporin 10; aquaporin-10; Small intestine aquaporin | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 89872 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96PS8 | |
| AQP10 | |
| Recombinant protein corresponding to Human Aquaporin 10 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Research Use Only