missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Aquaporin 1 Recombinant Rabbit Monoclonal Antibody (23GB1850)

Catalog No. MA554119
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA554119 20 μL
MA554118 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA554119 Supplier Invitrogen™ Supplier No. MA554119
Only null left

Rabbit Recombinant Monoclonal Antibody

Aquaporin 1 Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot

Aquaporin 1 is a 28kD integral membrane protein which was originally identified in red blood cells and renal proximal tubules. AQP1 is also expressed by the choroid plexus and various other tissues. It forms a water-specific channel that provides the plasma membranes of red cells and kidney proximal tubules with high permeability to water, thereby permitting water to move in the direction of an osmotic gradient.

Specifications

Antigen Aquaporin 1
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB1850
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Aqp1
Gene Accession No. P29972
Gene Alias AQP 1; AQP CHIP; Aqp1; AQP-1; AQP-CHIP; aquaporin 1; Aquaporin 1 (aquaporin channel forming integral protein 28 (CHIP)); aquaporin 1 (channel-forming integral protein, 28kDa, CO blood group); aquaporin 1 (Colton blood group); aquaporin 1, Colton blood group antigen; Aquaporin CHIP; Aquaporin1; aquaporin-1; Aquaporin-CHIP; channel-like integral membrane protein, 28-kDa; CHIP 28; CHIP28; CO; Colton blood group; Delayed early response protein 2; DER2; growth factor-induced delayed early response protein; membrane channel; MGC26324; urine water channel; Water channel protein for red blood cells and kidney proximal tubule
Gene Symbols Aqp1
Host Species Rabbit
Immunogen A synthesized peptide derived from human AQP1 (214-269AA).
Purification Method Affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 358
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less